Trequinsin hydrochloride ≥98% (durch HPLC)

Lieferant: Enzo Life Sciences

BMLPD1800010
ENZOBMLPD1800010EA 371 EUR
ENZOBMLPD1800010
Trequinsin hydrochloride ≥98% (durch HPLC)
Trequinsin hydrochloride

Ultrapotent, highly selective inhibitor of cGMP-inhibited phosphodiesterase (PDE lll, IC50=0.3nM) in vitro. Potentiates adenosine-stimulated cAMP accumulation.


  • PDE inhibitor


White to pale yellow solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml); slightly soluble in water.

Formel: C₂₄H₂₇N₃O₃·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD01076563
CAS-Nummer: 78416-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6dae48bccc6d2dc2a3f8ce84c9decd672385289&vid=5a779824d51a93a84a28a3d26d342a38e272d98f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR