Zardaverine ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
6-(4-(Difluoromethoxy)-3-methoxyphenyl)pyridazin-3(2H)-one
BMLPD1950005
ENZOBMLPD1950005EA
116
EUR
ENZOBMLPD1950005
Zardaverine ≥98% (durch HPLC)
Zardaverine
Selective, dual inhibitor of PDE III and PDE IV (IC50s=2.5 and 1.1 µM respectively). Anti-allergic activity, combined inhibition of PDE III and IV have an additive synergistic effect on inhibition of guinea pig bronchospasm.
- PDE inhibitor
Off-white to yellow solid.
Soluble in DMSO (25mg/ml).
Formel:
C₁₂H₁₀F₂N₂O₃ Molecular Weight: 268,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867059 CAS-Nummer: 101975-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...