BML-288

Lieferant: ENZO LIFE SCIENCES

BMLPD2000025
ENZOBMLPD2000025EA 402 EUR
ENZOBMLPD2000025
BML-288
BML-288

PDE2 inhibitor


Potent (IC50=40nM) and selective (290X selectivity over other PDE isoforms) inhibitor of phosphodiesterase Type II (PDE2). Displayed no significant level of binding to neither a collection of 54 receptors and ion channels, nor to a panel of 30 kinases. BML-288 demonstrated no inhibition of 5-lipoxygenase (5-LO) or cyclooxygenase (COX-1). BML-288 is more potent and selective than EHNA (BML-PD129).

Formel: C₁₄H₁₀N₄O₃S
Molecular Weight: 314,32 g/mol
CAS-Nummer: 851681-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe1ada877a7b60fe77037d7d10dbe442403b980a&vid=6f292a9fb1c7ceacce10d22a67e7fc0bb0e48b8b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR