Prostaglandin A1 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
PGA1, 9-oxo-15S-hydroxy-prosta-10,13E-dien-1-oic acid
BMLPG0010010
ENZOBMLPG0010010EA
279
EUR
ENZOBMLPG0010010
Prostaglandin A1 ≥98% (durch HPLC)
Prostaglandin A1
Biosynthesis from dihomo-γ-linolenic acid. Stimulates renin release. Antitumor activity. Inhibits NFκB activation in several human cell lines. Induces the synthesis of heat shock proteins. Activates PPARs in human osteosarcoma cells at 10μM.
- Prostanoid receptor ligand
White solid.
Soluble in 100% ethanol (50mg/ml), DMSO (50mg/ml) or PBS pH7.2 (2.4mg/ml).
Formel:
C₂₀H₃₂O₄ Molecular Weight: 336,5 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
14152-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...