Prostaglandin A2 ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Medullin, PGA2, Prosta-5,10,13-trien-1-oic acid, 15-hydroxy-9-oxo-, (5Z,13E,15S)-

BMLPG0020010
ENZOBMLPG0020010EA 313 EUR
ENZOBMLPG0020010
Prostaglandin A2 ≥99% (durch TLC)
Prostaglandin A2

Blocks cell cycle at the G1 and G2/M phase [1] by inhibiting G1 phase cyclin dependent kinases [2] downregulating cyclin D1 [3]. Modulates all three aspects of the glutathione-mediated biotransformation system, i.e. GSH levels, glutathione S-transferase activity and transport of GSH conjugates [4]. Antitumor activity. Induces γ-glutamylcysteine synthetase and apoptosis in L-1210 cells.


  • CDK inhibitor


Neat oil.


Soluble in 100% ethanol, acetone or ethyl acetate.

Formel: C₂₀H₃₀O₄
Molecular Weight: 334,5 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 13345-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR