Prostaglandin D₂ ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

BMLPG0050010
ENZOBMLPG0050010EA 465 EUR
ENZOBMLPG0050010
Prostaglandin D₂ ≥98% (durch HPLC)
Prostaglandin D2

Prostanoid receptor ligand.


Principal cyclooxygenase product of the mast cell and PGJ₂.

Lagertemperatur: Gefrierschrank
CAS-Nummer: 41598-07-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6bf1c5ec762d1895f102b42d464097a1ac5b08&vid=49761997255650b0123af013b9e17203b14721fb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR