Prostaglandin E1 ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: Alprostadil, Caverject, PGE1, 7-((1R,2R,3R)-3-Hydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)-5-oxocyclopentyl)heptanoic acid

BMLPG0060010
ENZOBMLPG0060010EA 197 EUR
ENZOBMLPG0060010
Prostaglandin E1 ≥99%
Prostaglandin E1

Prostanoid receptor ligand


Biosynthesis from dihomo-gamma-linolenic acid. A major prostaglandin in semen. Inhibits human platelet aggregation. Effects induced by receptor-mediated elevation of cAMP.

Formel: C₂₀H₃₄O₅
Molecular Weight: 354,487 g/mol
MDL: MFCD00077860
CAS-Nummer: 745-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fe98bd916cbc15cf02e6a5fbfda75468c680737&vid=ff7a79a75940284e49eda346ea73b0c9cac219d3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR