E-64-C
Lieferant: ENZO LIFE SCIENCES
BMLPI1060005
ENZOBMLPI1060005EA
371
EUR
ENZOBMLPI1060005
E-64-C
Proteine und Peptide
Calpain and cathepsin inhibitor
Cell-impermeable epoxysuccinyl peptide inhibitor of calpain and other cysteine proteases. Inhibits calpain and cathepsins B, H, and L.