E-64-C
Lieferant: ENZO LIFE SCIENCES
BMLPI1060005
ENZOBMLPI1060005EA 371 EUR
ENZOBMLPI1060005
E-64-C
Proteine und Peptide
Calpain and cathepsin inhibitor
Cell-impermeable epoxysuccinyl peptide inhibitor of calpain and other cysteine proteases. Inhibits calpain and cathepsins B, H, and L.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7f55ec737416e83220b9f24cf2090c03ce6e974&vid=88f654ab1c4b714a414de661af83e990b4972f3c&prid=web&clid=SAPDAM