Z-Leu-Leu-Leu-B(OH)2
Lieferant: ENZO LIFE SCIENCES
BMLPI1090100
ENZOBMLPI1090100EA 308 EUR
ENZOBMLPI1090100
Z-Leu-Leu-Leu-B(OH)2
Proteine und Peptide
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0,023 nM).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc8da17e53ac76b3fe9458e62c1ea69a8814ff8&vid=5678784526fd30b3df32719383c163e58a7b2096&prid=web&clid=SAPDAM