Z-Leu-Leu-Leu-B(OH)2
Lieferant: ENZO LIFE SCIENCES
BMLPI1090100
ENZOBMLPI1090100EA
308
EUR
ENZOBMLPI1090100
Z-Leu-Leu-Leu-B(OH)2
Proteine und Peptide
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0,023 nM).