DL-Thiorphan ≥97% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
2-(2-Benzyl-3-mercaptopropanamido)acetic acid
BMLPI1130005
ENZOBMLPI1130005EA
119
EUR
ENZOBMLPI1130005
DL-Thiorphan ≥97% (durch TLC)
DL-Thiorphan
Inhibits the metalloprotease neprilysin (neutral endopeptidase 24.11) and neprilysin 2. Inhibits opioid-degrading enzymes, potentiating THC-induced antinociception. Potentiates the effects of natriuretic peptides. Blocks enzymatic degradation of luteinizing hormone releasing hormone (LHRH).
- Neprilysin inhibitor
White powder.
Formel:
C₁₂H₁₅NO₃S Molecular Weight: 253.316 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058435 CAS-Nummer: 76721-89-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...