(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride ≥95% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
TLCK, Nalpha-p-Tosyl-L-lysine chloromethyl ketone hydrochloride, Tos-Lys-chloromethylketone·HCl
Inhibitor of trypsin and trypsin-like proteases
Irreversible inhibitor of trypsin and trypsin-like serine proteases such as granzyme D and tryptase. Also inhibits other proteases including plasmin, thrombin, and papain and some kinases including PKC. Inhibits LPS-induced expression of COX-2 in macrophages. Inhibits apoptosis (possibly through the inhibition of serine proteases). Chymotrypsin is not inhibited by TLCK, so this inhibitor can be used to block contaminating trypsin activity in chymoptrypsin preparations. TLCK can be used in combination with a chymotrypsin inhibitor to prevent proteolytic degradation during the isolation of proteins.
Formel:
C₁₄H₂₂Cl₂N₂O₃S Molecular Weight: 369,301 g/mol |
MDL:
MFCD00065395 CAS-Nummer: 4272-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...