N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, N-Tosyl-L-phenylalanyl Chloromethyl Ketone, (S)-N-(4-Chloro-3-oxo-1-phenyl-2-butyl)-p-toluenesulphonamide, TPCK

BMLPI1220001
ENZOBMLPI1220001EA 211 EUR
ENZOBMLPI1220001
N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone ≥98%
N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone

Proteasome inhibitor


Irreversible inhibitor of chymotrypsin-like serine proteases such as granzyme D. Inhibits LPS-induced expression of COX-2 in macrophages. Inhibits γ-irradiation-induced apoptosis in myeloid leukemic cells.

Formel: C₁₇H₁₈ClNO₃S
Molecular Weight: 351,85 g/mol
MDL: MFCD00000935
CAS-Nummer: 402-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbbc647c19ba4676799b97b11ba7148c3e8427ca&vid=e485d9643b20b3c34843f5ed983a6f8d8d39b0e4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR