NapSul-Ile-Trp-CHO
Lieferant: ENZO LIFE SCIENCES
BMLPI1250005
ENZOBMLPI1250005EA
325
EUR
ENZOBMLPI1250005
NapSul-Ile-Trp-CHO
Proteine und Peptide
Cathepsin inhibitor
Potent and selective inhibitor of cathepsin L, a lysosomal cysteine protease. IC50=1,9 nM.