NapSul-Ile-Trp-CHO
Lieferant: ENZO LIFE SCIENCES
BMLPI1250005
ENZOBMLPI1250005EA 325 EUR
ENZOBMLPI1250005
NapSul-Ile-Trp-CHO
Proteine und Peptide
Cathepsin inhibitor
Potent and selective inhibitor of cathepsin L, a lysosomal cysteine protease. IC50=1,9 nM.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81379cf8629e1830e76d669f70b4e22aefd0d415&vid=43eadeae21737dc9d5e8e4fb2dec6b47df11667b&prid=web&clid=SAPDAM