STT negative control for gelatinase
Lieferant: ENZO LIFE SCIENCES
BMLPI1370005
ENZOBMLPI1370005EA 385 EUR
ENZOBMLPI1370005
STT negative control for gelatinase
Proteine und Peptide
MMP inhibitor
STTHWGFTLS is ten times less potent than, thus a negative control for CTTHWGFTLC (Prod. No. BML-PI136), which is a specific inhibitor of the gelatinases MMP-2 and MMP-9. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo.