Apstatin ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutanoyl]-L-prolyl-L-prolyl-L-alaninamide
BMLPI1450001
ENZOBMLPI1450001EA
165
EUR
ENZOBMLPI1450001
Apstatin ≥95% (durch HPLC)
Apstatin
Apstatin is a potent and selective inhibitor of aminopeptidase P (APP), Ki=2.6 µM for purified rat lung membrane-bound APP. It blocks the APP-mediated degradation of bradykinin. Limits myocardial infarct size alone or with ACE inhibitors.
- Aminopeptidase inhibitor
Solid.
Soluble in water (>25 mg/ml) or DMSO (20 mg/ml).
Formel:
C₂₃H₃₃N₅O₅ Molecular Weight: 459,5 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
160470-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...