BML-260 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLPR1190010
ENZOBMLPR1190010EA
91.3
EUR
ENZOBMLPR1190010
BML-260 ≥98% (durch TLC)
BML-260
A potent inhibitor of the dual specificity phosphatase JSP-1 (IC50=18µM).
- JSP-1 inhibitor
Yellow solid.
Soluble in DMSO (25mg/ml).
Formel:
C₁₇H₁₁NO₃S₂ Molecular Weight: 341,4 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
101439-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...