BML-267
Lieferant: ENZO LIFE SCIENCES
BMLPR1210050
ENZOBMLPR1210050EA 590 EUR
ENZOBMLPR1210050
BML-267
BML-267
PTP1B inhibitor
Potent and selective protein tyrosine phosphatase 1B (PTP1B) inhibitor (IC50=11 µM).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b665585d628af7ea580ed6b748c1f9e6e1a3079a&vid=0e1977d6b9123ae9c055be1ff2fea3bd8193fa3a&prid=web&clid=SAPDAM