BML-267
Lieferant: ENZO LIFE SCIENCES
BMLPR1210050
ENZOBMLPR1210050EA
590
EUR
ENZOBMLPR1210050
BML-267
BML-267
PTP1B inhibitor
Potent and selective protein tyrosine phosphatase 1B (PTP1B) inhibitor (IC50=11 µM).