BMOV ≥96% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Bis(maltolato)oxovanadium (IV)
BMLPR1310050
ENZOBMLPR1310050EA
84.7
EUR
ENZOBMLPR1310050
BMOV ≥96% (durch TLC)
BMOV
Organic vanadium complex which acts as a potent insulin mimic.
Soluble in water (10 mg/ml) and DMSO (15 mg/ml).
Formel:
C₁₂H₁₀O₇V Lagertemperatur: Raumtemperatur |
CAS-Nummer:
38213-69-3 |
Spezifikation Testergebnisse
Purity | ≥96% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...