Proteasome ELISA Kit
Lieferant: ENZO LIFE SCIENCES
This kit provides the means to quantify 20S proteasome concentrations in biological samples using a sandwich ELISA technique, utilizing two 20S proteasome specific antibodies for capture and detection purposes together with a highly sensitive substrate. Sample 20S proteasome levels are determined by comparison to a 20S proteasome calibration curve produced in parallel. This kit provides sufficient material for 1x96 well plate set-up to be run.
- Determination of 20S proteasome levels in biological samples (cell lysates, tissue extracts, plasma, serum)
- Comparison of 20S proteasome levels in plasma/serum samples associated with a particular disease/illness with samples from healthy controls
- Investigation of variation in 20S proteasome levels in abnormal cell lines/tissues
- Assay Time: <3.5 hours
Lieferumfang: Kit consists of 96 wells (24 tests in triplicate), plate, capture antibody, detection antibody, conjugated antibody, Proteasome stock solution, binding buffer, wash buffer, blocking buffer, ELISA buffer, lysis buffer, TMB substrate solution, stop solution.