MK-571 sodium salt 90 HPLC_ASSAY_METHOD

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: Sodium (E)-3-(((3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)((3-(dimethylamino)-3-oxopropyl)thio)methyl)thio)propanoate, MK-571 sodium

BMLRA1090005 BMLRA1090025
ENZOBMLRA1090005EA 86.9 EUR
ENZOBMLRA1090005 ENZOBMLRA1090025
MK-571 sodium salt 90 HPLC_ASSAY_METHOD
MK-571 sodium salt

A potent, selective, orally active leukotriene D₄ antagonist. Ki=2.1 nM for inhibition of [³H]-LTD₄ binding to human lung membranes. Competitively antagonizes contractions of guinea pig trachea and ileum and human trachea induced by LTD₄. Blocks LTD₄ and ascaris-induced bronchoconstriction in conscious squirrel monkeys.


Soluble in water (20 mg/ml), aqueous buffers, 100% ethanol (10 mg/ml) or DMSO.

Formel: C₂₆H₂₆ClN₂O₃S₂·Na
Molecular Weight: 537 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11114392
CAS-Nummer: 115103-85-0

Bestellen

Spezifikation Testergebnisse

Purity ≥90% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR