Latanoprost ≥98%

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: Latanoprost, (Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R)-3-hydroxy-5-phenylpentyl)cyclopentyl)hept-5-enoate

BMLRA1200005
ENZOBMLRA1200005EA 324 EUR
ENZOBMLRA1200005
Latanoprost ≥98%
Latanoprost

Prostanoid receptor ligand.


Prodrug (isopropyl ester) of 17-phenyl-13,14-dihydro-prostaglandin F₂α. The free acid form is a potent agonist at FP receptors. It is in clinical use as a topical antiglaucoma agent.

Formel: C₂₆H₄₀O₅
Molecular Weight: 432.60 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00216074
CAS-Nummer: 130209-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27906802b7f372f45e5995778c5f03f499edd955&vid=658e763884b93a19eeaa6b7082f36a2ab211bbfd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (Assay)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR