Deoxyfuconojirimycin hydrochloride ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (2S,3R,4S,5R)-2-Methyl-3,4,5-trihydroxypiperidine hydrochloride

BMLS1080005
ENZOBMLS1080005EA 180 EUR
ENZOBMLS1080005
Deoxyfuconojirimycin hydrochloride ≥98% (durch TLC)
Deoxyfuconojirimycin hydrochloride

A potent and selective inhibitor of α-L-fucosidases.


  • Off-white solid


Soluble in water

Formel: C₆H₁₃NO₃·HCl
Molecular Weight: 183,6 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 210174-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb27805e6745eb3dbe7cc04603d83e95b758d89&vid=07e6d957012b300d52356b07dfa2d5f8e243bf8c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR