Bovine native Calmodulin (from Brain), Agarose
Lieferant: Enzo Life Sciences
BMLSE1020001
ENZOBMLSE1020001EA 681 EUR
ENZOBMLSE1020001
Bovine native Calmodulin (from Brain), Agarose
Proteine und Peptide
Isolated from bovine brain. Calmodulin conjugated to 4% cross-linked agarose beads via a 5 atom hydrophilic spacer arm.