Human EGF receptor
Lieferant: ENZO LIFE SCIENCES
BMLSE1160010
ENZOBMLSE1160010EA
608
EUR
ENZOBMLSE1160010
Human EGF receptor
Proteine und Peptide
Isolated from human A431 cells.
EGFR is active but not autophosphorylated.