Human recombinant Casein kinase II (from E. coli)
Lieferant: ENZO LIFE SCIENCES
BMLSE1240010
ENZOBMLSE1240010EA 416 EUR
ENZOBMLSE1240010
Human recombinant Casein kinase II (from E. coli)
Enzyme
Produced in E. coli. Consists of 44 kDa alpha catalytic and 26 kDa & szlig; regulatory subunits as an alpha2& szlig;2 tetramer.