Human recombinant Caspase-3 (from E. coli)
Lieferant: ENZO LIFE SCIENCES
BMLSE1695000
ENZOBMLSE1695000EA
516
EUR
ENZOBMLSE1695000
Human recombinant Caspase-3 (from E. coli)
Enzyme
Produced in E. coli. The enzyme is cleaved and activated from the proenzyme.
Caspase-3 (CPP32; Yama; apopain) from human cDNA, expressed in an E. coli expression system. Can be used to study enzyme regulation and kinetics, cleave target substrates and screen for inhibitors.