Human recombinant Caspase-7 (from E. coli)
Lieferant: ENZO LIFE SCIENCES
BMLSE1775000
ENZOBMLSE1775000EA
555
EUR
ENZOBMLSE1775000
Human recombinant Caspase-7 (from E. coli)
Enzyme
Produced in E. coli. Human caspase-7 (aa 24-303). Genbank Accession No. U40281.