Human recombinant Caspase-9 (from E. coli)
Lieferant: ENZO LIFE SCIENCES
BMLSE2400050
ENZOBMLSE2400050EA 516 EUR
ENZOBMLSE2400050
Human recombinant Caspase-9 (from E. coli)
Enzyme
Produced in E. coli. Full length human caspase-9 (aa 1-416).