Human Neutrophil elastase (from neutrophils)
Lieferant: ENZO LIFE SCIENCES
BMLSE2840100
ENZOBMLSE2840100EA
459
EUR
ENZOBMLSE2840100
Human Neutrophil elastase (from neutrophils)
Enzyme
Isolated from human neutrophils. Active enzyme.
- Functions in inflammation
Can cleave a variety of proteins, including fibrous elastin, collagen, clotting factors, TNF-α , and α 1- antichymotrypsin. Because of its potential roles in inflammatory diseases such as emphysema and cystic fibrosis, it is a target for drug discovery.