Human Recombinant 14-3-3zeta (GST-tag) (from E. coli), GST Tag
Lieferant: Enzo Life Sciences
BMLSE3180100
ENZOBMLSE3180100EA 374 EUR
ENZOBMLSE3180100
Human Recombinant 14-3-3zeta (GST-tag) (from E. coli), GST Tag
Proteine und Peptide
Produced in E. coli. Human 14-3-3ζ is fused at the N-terminus to a GST-tag.