Human recombinant p70 S6 Kinase
Lieferant: ENZO LIFE SCIENCES
BMLSE3450005
ENZOBMLSE3450005EA
327
EUR
ENZOBMLSE3450005
Human recombinant p70 S6 Kinase
Enzyme
Produced in insect cells. Active, full length p70 S6K.