Human recombinant PKCν (GST-tag),GST tag
Lieferant: ENZO LIFE SCIENCES
BMLSE3490005
ENZOBMLSE3490005EA
280
EUR
ENZOBMLSE3490005
Human recombinant PKCν (GST-tag),GST tag
Proteine und Peptide
Produced in insect cells. Active, full length protein kinase C nu (PKCν, protein kinase D3, PKD3) is fused at the N-terminus to a GST-tag.