Human thrombin (from plasma)
Lieferant: ENZO LIFE SCIENCES
BMLSE3631000
ENZOBMLSE3631000EA
516
EUR
ENZOBMLSE3631000
Human thrombin (from plasma)
Enzyme
Isolated from human plasma as prothrombin, and activated with Factor Xa, Factor Va, phospholipids, and calcium to yield a-thrombin.