Human recombinant Lyn B (GST-tag),GST Tag
Lieferant: ENZO LIFE SCIENCES
BMLSE3850005
ENZOBMLSE3850005EA 327 EUR
ENZOBMLSE3850005
Human recombinant Lyn B (GST-tag),GST Tag
Proteine und Peptide
Produced in insect cells. Full length active enzyme.