Human recombinant FAP
Lieferant: ENZO LIFE SCIENCES
BMLSE4090010
ENZOBMLSE4090010EA 687 EUR
ENZOBMLSE4090010
Human recombinant FAP
Proteine und Peptide
Produced in a baculovirus expression system. Active recombinant human FAP (aa 29-760), representing a naturally-occurring cleaved (soluble) form.