Human recombinant PI3 kinase p110 alpha/p85 alpha
Lieferant: ENZO LIFE SCIENCES
BMLSE4360020
ENZOBMLSE4360020EA
714
EUR
ENZOBMLSE4360020
Human recombinant PI3 kinase p110 alpha/p85 alpha
Enzyme
Produced in insect cells. Complex of human full length p110a and p85a subunits of phosphatidylinositol 3-kinase.