Human Recombinant 14-3-3gamma (from E. coli), HIS Tag
Lieferant: ENZO LIFE SCIENCES
BMLSE4820100
ENZOBMLSE4820100EA 400 EUR
ENZOBMLSE4820100
Human Recombinant 14-3-3gamma (from E. coli), HIS Tag
Proteine und Peptide
Produced in E. coli. Human 14-3-3gamma is fused at the C-terminus to a His-tag.