Human recombinant DPP9
Lieferant: ENZO LIFE SCIENCES
BMLSE5280010
ENZOBMLSE5280010EA
710
EUR
ENZOBMLSE5280010
Human recombinant DPP9
Proteine und Peptide
Produced in insect cells. Active recombinant human DPP9 with an N-terminal purification tag.