Human recombinant BACE2 (soluble),HIS Tag
Lieferant: ENZO LIFE SCIENCES
BMLSE5500010
ENZOBMLSE5500010EA 473 EUR
ENZOBMLSE5500010
Human recombinant BACE2 (soluble),HIS Tag
Proteine und Peptide
Highly active
Produced in insect cells.
Recombinant soluble BACE2, cloned from human cDNA, secreted as zymogen from insect cells, purified using a C-terminal His-tag, and cleaved to the mature active form. BACE2 is an aspartic protease found in both transmembrane and soluble forms. It is related to, and possesses an overlapping substrate profile with, BACE1. It is likely involved in Alzheimer’s disease, and implicated in other neurological, pancreatic, and muscle pathologies.