C2-Dihydroceramide ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLSL1010005
ENZOBMLSL1010005EA 86.9 EUR
ENZOBMLSL1010005
C2-Dihydroceramide ≥98% (durch TLC)
C2-Dihydroceramide

Negative control for C2 ceramide.


C₂ Dihydroceramide is inactive and may be used as a negative control for C₂ ceramide.


Soluble in ethanol (5 mg/ml) or DMSO (5 mg/ml: dissolve in hot DMSO, then cool to RT).

Formel: C₂₀H₄₁NO₃
Molecular Weight: 343,552 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00236495
CAS-Nummer: 13031-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b9c079541e2389f96c2d8226a0b48f96233b51c&vid=8f87bed7043a0f3124e5773f3847f1034e1e01a6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR