Dihydroceramide C6 ≥98%

Lieferant: ENZO LIFE SCIENCES

BMLSL1110005
ENZOBMLSL1110005EA 84.7 EUR
ENZOBMLSL1110005
Dihydroceramide C6 ≥98%
Dihydroceramide C6

Negative control for C6 ceramide.


C₆ Dihydroceramide is inactive and may be used as a negative control for C₆ ceramide.


Soluble in DMF (25 mg/ml), DMSO (5 mg/ml) or ethanol (10 mg/ml).

Formel: C₂₄H₄₉NO₃
Molecular Weight: 399.66 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01320384
CAS-Nummer: 171039-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b578ea78f88b6da551367e6e57c7bf3cf13864fa&vid=ec4552e34061f9fc5fb856ac2c5eead9504e8041&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR