Dihydroceramide C8 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLSL1130025
ENZOBMLSL1130025EA 384 EUR
ENZOBMLSL1130025
Dihydroceramide C8 ≥98% (durch TLC)
Dihydroceramide C8

Negative control for C8 ceramide.


C₈ Dihydroceramide is inactive and may be used as a negative control for C₈ ceramide.


Soluble in DMF (25 mg/ml), DMSO (5 mg/ml) or ethanol (10 mg/ml).

Formel: C₂₆H₅₃NO₃
Molecular Weight: 427.714 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01320385
CAS-Nummer: 145774-33-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9c51dab8b79a506c7159aafae05ad9a664bf2d9&vid=a9f8fe2b08db666876652f470fa98df08c6a4636&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR