D-erythro-C8 Ceramine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLSL1450005
ENZOBMLSL1450005EA 384 EUR
ENZOBMLSL1450005
D-erythro-C8 Ceramine ≥98% (durch TLC)
D-erythro-C8-ceramine

Ceramide-activated protein kinase activator.


C₈ ceramine is an analog of ceramide in which the amide is substituted with an amine. This substitution renders it inert to ceramidases. It is highly effective at induction of apoptosis in U937 cells. Ceramine induces peak DNA fragmentation in 6 hours compared to 18 hours for C₈ ceramide.


Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml).

Formel: C₂₆H₅₃NO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD01632747
CAS-Nummer: 170926-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=605725c70b415e54491c0f1a364bf7a374e4a1cd&vid=bd1d7d6943122cbce51103ccdacaaafcd6a89dfc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR