Phytoceramide C2 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLSL1510005
ENZOBMLSL1510005EA 348 EUR
ENZOBMLSL1510005
Phytoceramide C2 ≥98% (durch TLC)
Phytoceramide C2

Inhibits yeast growth and activates ceramide-activated protein phosphatase (CAPP).


  • Ceramide-activated protein phosphatase activator


White to off-white solid.


Soluble in ethanol (>25 mg/ml) or DMSO (>25 mg/ml with warming).

Formel: C₂₀H₄₁NO₄
Molecular Weight: 359,551 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02686954
CAS-Nummer: 475995-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0af7ccf85f1a2f330155f55a9cc4fa0cedb86376&vid=63d0b050b5fa75f2d4109fbe1014152a72be23e4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR