L-erythro-MAPP ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLSL2220025
ENZOBMLSL2220025EA 239 EUR
ENZOBMLSL2220025
L-erythro-MAPP ≥98% (durch TLC)
L-erythro-MAPP

Negative control for D-erythro-MAPP


Useful as a negative control for D-erythro-MAPP. It does not raise endogenous ceramide levels or inhibit the growth of HL-60 cells.


Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).

Formel: C₂₃H₃₉NO₂
Molecular Weight: 361,6 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 143492-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bf6e28f293b71bdec92483da91cd4daf691dc8d&vid=9eb02c0586a4610fe9b3cab0068f7a4f2f517891&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR