Tricyclodecan-9-yl xanthogenate potassium salt ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLST3300005 BMLST3300025
ENZOBMLST3300005EA 86.9 EUR
ENZOBMLST3300005 ENZOBMLST3300025
Tricyclodecan-9-yl xanthogenate potassium salt ≥98% (durch TLC)
Tricyclodecan-9-yl xanthogenate potassium salt

Selective inhibitor of phosphatidylcholine-specific phospholipase C (PC-PLC). Has been used to study the coupling of phosphatidylcholine-specific phospholipase C with sphingomyelinase. Shows antitumor and antiviral activity. Inhibits basement membrane collagen synthesis, which is the last step in the development of a new blood vessel. Inhibits induction of nitric oxide synthases (NOS). Induces apoptosis. It indirectly inhibits phorbol ester stimulated PLD activity.


  • Phospholipase inhibitor


Off-white solid.


Soluble in water (25mg/ml).

Formel: C₁₁H₁₅KOS₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00171401
CAS-Nummer: 83373-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR