BML-280 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: VU0285655-1, N-(2-(4-Oxo-1-phenyl-1,3,8-triazaspiro[4.5]decan-8-yl)-ethyl)quinoline-3-carboxamide

BMLST3710005
ENZOBMLST3710005EA 110 EUR
ENZOBMLST3710005
BML-280 ≥98% (durch TLC)
BML-280

Potent and selective inhibitor of phospholipase D2 (PLD2) in cellular (PLD1 IC50 = 1900nM Calu-1 cells; PLD2 IC50 = 90nM HEK293-gfpPLD2 cells) and in vitro (hPLD1 IC50 = >20’000nM; hPLD2 IC50 = 500nM) assays.


  • PLD inhibitor


Off-white solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).

Formel: C₂₅H₂₇N₅O₂
Molecular Weight: 429,5 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 1158347-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=812a5346a907e520a5336543c69f563d42b614ec&vid=22b7d78bc5c0f09fcaf1c169e83892e26ea9884e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR