BML-279
Lieferant: ENZO LIFE SCIENCES
BMLST3720005
ENZOBMLST3720005EA 110 EUR
ENZOBMLST3720005
BML-279
BML-279
PLD inhibitor.
Dual phospholipase D1/D2 (PLD1/PLD2) inhibitor in cellular assays (PLD1 IC50=8 nm in Calu-1 cells; PLD2 IC50=42 nm in HEK293-gfpPLD2 cells).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9653dbb493f0caa680f880bc9d3acb315ff03f91&vid=dd09b98711e59565152ca7f045d60b28567fb0e7&prid=web&clid=SAPDAM