(±)-Propranolol hydrochloride ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, Propranolol hydrochloride, DL-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride

BMLST4050005
ENZOBMLST4050005EA 67.1 EUR
ENZOBMLST4050005
(±)-Propranolol hydrochloride ≥98% (durch HPLC)
(±)-Propranolol hydrochloride

Propranolol blocks PLD derived DAG formation by inhibiting phosphatidate phosphohydrolase (IC50=70 µM). It is a useful tool to discriminate between PLC or PLD mediated DAG production. It is also a β-adrenergic receptor blocking agent.


  • Adrenergic receptor inhibitor


White to off-white powder.


Soluble in water.

Formel: C₁₆H₂₁NO₂·HCl
Molecular Weight: 295,81 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012558
CAS-Nummer: 318-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR