2-Morpholino-8-phenylchromone ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLST4200005 BMLST4200025
ENZOBMLST4200005EA 165 EUR
ENZOBMLST4200005 ENZOBMLST4200025
2-Morpholino-8-phenylchromone ≥98% (durch TLC)
2-Morpholino-8-phenylchromone

PI3 kinase inhibitor


Potent, cell permeable, inhibitor of PI(3)K (phosphoinositide 3-kinase) that acts on the ATP-binding site of the enzyme. Displays an IC50 of 1.4 µM and inhibits all isoforms equally in in vitro assays using purified PI 3-kinase. (p110alpha IC50=0.63µM)Inhibits Pim-1 kinase with IC50=4.0µM.Other kinases such as PKC, PKA, MAP kinase, S6 kinase, EGF receptor tyrosine kinase, c-src kinase, PI 4-kinase, diacylglycerol kinase or rabbit kidney ATPase are not inhibited at 50µM. Induces apoptosis in many cell types by blocking the PI3-K/Akt anti-apoptotic pathway. Useful tool for identifying cellular events which are regulated by the PI3-K/Akt axis. Sensitizes tumor cells to drug-induced apoptosis independent of its PI3K inhibitory activity. For a negative control see LY-303,511 (Prod. No. ALX-270-410).

Formel: C₁₉H₁₇NO₃
MDL: MFCD00270881
CAS-Nummer: 154447-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87e4d94853fe8cc98d8f504b791c41d538783557&vid=87b1286d5a0b14660e58ae7745090d6c281c62a4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR